Molybden, gauze woven from wire, twilled weave 165 mesh, Ø 0.0509 mm (0.002 in)
Leverandør: Thermo Fisher Scientific
42404.FL 42404.GT
42404.FLEA 3725 NOK
42404.FL 42404.GT
Molybden, gauze woven from wire, twilled weave 165 mesh, Ø 0.0509 mm (0.002 in)
Molybden
CAS No.: 7439-98-7 Physical Form: Wire Cloth MDL No.: MFCD00003465
Formel: Mo
Kokepunkt: 3617 °C (1013 hPa)
Smeltepunkt: 2622 °C
Tetthet: 10,28 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003465
CAS-nummer: 7439-98-7
EINECS: 231-107-2
Merck Indeks: 12,06317