Kobalt ≥99.8% (metals basis), spherical powder -100/+325 mesh
Leverandør: Thermo Scientific
Danger
41605.18 41605.30
41605.18EA 1825 NOK
41605.18 41605.30
Kobalt ≥99.8% (metals basis), spherical powder -100/+325 mesh
Kobalt
Purity: 99.8% CAS No.: 7440-48-4 Appearance: Black Odour: Odourless UN No.: 3089 MDL No.: MFCD00010935
Formel: Co
MW: 58,93 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010935
CAS-nummer: 7440-48-4
EINECS: 231-158-0
FN: 3089
ADR: 4.1,III