Nikkel ≥99.8% (metals basis), spherical powder, APS 5-15 µm
Leverandør: Thermo Scientific
Danger
00663.14 00663.22
00663.14EA 1175 NOK
00663.14 00663.22
Nikkel ≥99.8% (metals basis), spherical powder, APS 5-15 µm
Nikkel
Purity: 99.8% CAS No.: 7440-02-0 Physical Form: Apparent density 3-5g/cm3 Odour: Odourless UN No.: 3089 MDL No.: MFCD00011137
Formel: Ni
MW: 58,69 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011137
CAS-nummer: 7440-02-0
EINECS: 231-111-4
FN: 3089
ADR: 4.1,II