Nikkel ≥99.95% (metals basis), thin foil, Thickness: 0.01 mm (0.0004 in)
Leverandør: Thermo Fisher Scientific
Danger
35817.FF 35817.FI 35817.GH
35817.FFEA 800 NOK
35817.FF 35817.FI 35817.GH
Nikkel ≥99.95% (metals basis), thin foil, Thickness: 0.01 mm (0.0004 in)
Nikkel
Purity: 99.95% CAS No.: 7440-02-0 Physical Form: ≈0.055g/25x25mm, not certified pinhole free Odour: Odourless MDL No.: MFCD00011137
Formel: Ni
MW: 58.69 g/mol
Kokepunkt: 3007
Smeltepunkt: 1453
Tetthet: 8.9 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011137
CAS-nummer: 7440-02-0
EINECS: 231-111-4
FN: 3089
ADR: 4.1 III
Merck Indeks: 12,06582