D(+)-Glukose monohydrat for microbiology pH 6 - 7 (100 g/l, H₂O, 20 °C), Millipore®
Leverandør: Merck
M-Clarity™ quality level = MQ100
1.08342.9025EA 0 NOK
1.08342.9010 1.08342.9025
D(+)-Glukose monohydrat for microbiology pH 6 - 7 (100 g/l, H₂O, 20 °C), Millipore®
D(+)-Glukose monohydrat
Formel: C₆H₁₂O₆·H₂O
MW: 198,17 g/mol
Smeltepunkt: 83 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149450
CAS-nummer: 14431-43-7
EINECS: 200-075-1
https://no.vwr.com/stibo/web/std.lang.all/21/23/2302123.jpg