DL-Tyrosine 99%
Leverandør: Thermo Scientific
Warning
Synonymer: 4-Hydroxyphenylalanine, 3-(p-Hidroxifenil)-DL-alanina, H-DL-Tyr-OH
163570250 163571000
ACRO163570250EA 1100 NOK
ACRO163570250 ACRO163571000
DL-Tyrosine 99%
DL-Tyrosine
Formel: C₉H₁₁NO₃
MW: 181,19 g/mol
Kokepunkt: 378 °C (1013 hPa)
Smeltepunkt: 325 °C (dec.)
MDL nummer: MFCD00063074
CAS-nummer: 556-03-6
EINECS: 209-113-1
Merck Indeks: 12,09970
Bestill nå
Spesifikasjon Testresultater
Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with HClO4 98.5 to 101.5 %
NMR (1H-NMR) Consistent with structure