2,3,5-Trifenyltetrazoliumklorid, white powder
Leverandør: MP Biomedicals
Warning
Synonymer: Tetrazolium salt, Tetrazolium Red, 2,3,5-Triphenyl-2H-tetrazol-3-ium chloride, 2,3,5-Triphenyl-2H-tetrazolium chloride, TTC
0210312610 0210312625 0210312650 0210312680
ICNA0210312610EA 930 NOK
ICNA0210312610 ICNA0210312625 ICNA0210312650 ICNA0210312680
2,3,5-Trifenyltetrazoliumklorid, white powder
2,3,5-Trifenyltetrazoliumklorid
2,3,5-Triphenyltetrazolium Chloride is a redox indicator. In the TTC assay (also known as TTC test or tetrazolium test), TTC is used to differentiate between metabolically active and inactive tissues. The white compound is enzymatically reduced to red TPF (1,3,5-triphenylformazan) in living tissues due to the activity of various dehydrogenases (enzymes important in oxidation of organic compounds and thus cellular metabolism), while it remains as white TTC in areas of necrosis since these enzymes have been either denatured or degraded.
2,3,5-Triphenyltetrazolium Chloride is commonly used in biochemical experiments especially to indicate cellular respiration. TTC has been employed in autopsy pathology to assist identification of post-mortem myocardial infarctions. Healthy viable heart muscle will stain deep red from the cardiac lactate dehydrogenase; while areas of potential infarctions will be more pale. Useful indicator for reducing substances
  • Presentation: White Powder
  • Soluble in ethanol, alcohol, water (10 mg/mL - clear, colorless solution) or acetone. Insoluble in ether.
  • Storage temperature: +4 °C, protect from light
2,3,5-Triphenyltetrazolium Chloride is a redox indicator. In the TTC assay (also known as TTC test or tetrazolium test), TTC is used to differentiate between metabolically active and inactive tissues. The white compound is enzymatically reduced to red TPF (1,3,5-triphenylformazan) in living tissues due to the activity of various dehydrogenases (enzymes important in oxidation of organic compounds and thus cellular metabolism), while it remains as white TTC in areas of necrosis since these enzymes have been either denatured or degraded.
2,3,5-Triphenyltetrazolium Chloride is commonly used in biochemical experiments especially to indicate cellular respiration. TTC has been employed in autopsy pathology to assist identification of post-mortem myocardial infarctions. Healthy viable heart muscle will stain deep red from the cardiac lactate dehydrogenase; while areas of potential infarctions will be more pale. Useful indicator for reducing substances
Formel: C₁₉H₁₅ClN₄
MW: 334,81 g/mol
Kokepunkt: ∼360 °C (1013 hPa)
Smeltepunkt: 243…250 °C (lit.)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00011963
CAS-nummer: 298-96-4
EINECS: 206-071-6
Merck Indeks: 13,09815
Bestill nå
Spesifikasjon Testresultater
Identity Test Passes
FTIR Conforms to Standard
I max (water) 246 ± 5 nm