Tris-(hydroksymetyl)aminometan hydroklorid, Millipore®
Leverandør: Merck Millipore Calbiochem
Synonymer: Tris(hydroxymethyl)methylamine hydrochloride, 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride, Trizma® hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl, Tromethane hydrochloride, Tris-HCl, Tris(hydroxymethyl)methylammonium chloride, Trometamol hydrochloride, THAM HCl
648317-1 648317-100
648317-1EA 8325 NOK
648317-1 648317-100
Tris-(hydroksymetyl)aminometan hydroklorid, Millipore®
Tris-(hydroksymetyl)aminometan hydroklorid
Formel: NH₂C(CH₂OH)₃·HCl
MW: 157,6 g/mol
Smeltepunkt: 150…152 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012590
CAS-nummer: 1185-53-1
EINECS: 214-684-5
Merck Indeks: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM