N-Acetyl-L-cystein, Millipore®
Leverandør: Merck
Warning
Synonymer: Acetylcysteine, N-Acetyl-3-mercaptoalanine, NAC, (R)-(+)-N-Acetylcysteine, (R)-2-Acetamido-3-mercaptopropanoic acid, Ac-Cys-OH, N-Acetyl-L-Cysteine, LNAC
M-Clarity™ quality level = MQ100
1.12422.0025 1.12422.0100 1.12422.1000 1.12422.9010
1.12422.0025EA 400 NOK
1.12422.0025 1.12422.0100 1.12422.1000 1.12422.9010
N-Acetyl-L-cystein, Millipore®
N-Acetyl-L-cysteine
Formel: C₅H₉NO₃S
MW: 163,2 g/mol
Kokepunkt: 371 °C (1013 hPa)
Smeltepunkt: 106…108 °C
Tetthet: 1,48 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004880
CAS-nummer: 616-91-1
EINECS: 210-498-3
Merck Indeks: 13,00090