1,10-Diaminodecane ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: Decane-1,10-diamine

D0085-25G D0085-100G
TCIAD0085-25GEA 1150 NOK
TCIAD0085-25G TCIAD0085-100G
1,10-Diaminodecane ≥98.0% (by GC, titration analysis)
1,10-Diaminodecane
Formel: C₁₀H₂₄N₂
MW: 172,31 g/mol
Kokepunkt: 140°C
Smeltepunkt: 60 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008151
CAS-nummer: 646-25-3
EINECS: 211-471-9
FN: 3259
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=418d664dc09033a9c1ac6acbbb90be03ba8b1019&vid=418d664dc09033a9c1ac6acbbb90be03ba8b1019&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 59.0 to 64.0 deg-C
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR