2-Ethylaminoethanol 98%

Leverandør: Thermo Scientific
Danger

Synonymer: N-Ethyl-2-aminoethanol, N-Ethyl-2-ethanolamin, 2-(Ethylamino)ethan-1-ol

168730010 168732500
ACRO168730010EA 895 NOK
ACRO168730010 ACRO168732500
2-Ethylaminoethanol 98%
2-Ethylaminoethanol
Formel: C₄H₁₁NO
MW: 89,14 g/mol
Kokepunkt: 167 °C (1013 hPa)
Smeltepunkt: –6 °C
Tetthet: 0,91 g/cm³
Flash punkt: 78 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002841
CAS-nummer: 110-73-6
EINECS: 203-797-5
FN: 2735
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4390 to 1.4430 (20°C, 589 nm)
Water ≤0.5 %
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR