Aluminiumtriisopropanolat 99.99% (trace metals basis)
Leverandør: Thermo Scientific
Danger
Synonymer: Aluminiumtriisopropylate, Aluminiumisopropylate, Aluminiumtriisopropoxide, Aluminium isopropoxide
212230100 212230500 212232500
ACRO212230100EA 1150 NOK
ACRO212230100 ACRO212230500 ACRO212232500
Aluminiumtriisopropanolat 99.99% (trace metals basis)
Aluminiumtriisopropanolat
Formel: C₉H₂₁AlO₃
MW: 204,25 g/mol
Kokepunkt: 125…130 °C (38 mmHg)
Smeltepunkt: 129…136 °C
Tetthet: 0,9771 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008870
CAS-nummer: 555-31-7
EINECS: 209-090-8
FN: 3181
ADR: 4.1,II
Merck Indeks: 13,00346
Bestill nå
Spesifikasjon Testresultater
Appearance White Powder and/or chunks
Infrared spectrum Conforms
Total trace metal impurities ≤150 ppm