Sodium-1-decanesulphonate 96%
Leverandør: Thermo Scientific
369450050 369450250 369451000
ACRO369450050EA 1025 NOK
ACRO369450050 ACRO369450250 ACRO369451000
Sodium-1-decanesulphonate 96%
Sodium-1-decanesulphonate
Formel: C₁₀H₂₁NaO₃S
MW: 244,33 g/mol
Smeltepunkt: >300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007526
CAS-nummer: 13419-61-9
EINECS: 236-525-9
Bestill nå
Spesifikasjon Testresultater
Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤5 % (100°C, 3 h) (vacuum)
Titration after Ion exchange ≥95 % (On dry substance)