2,2-Dimetyl-1-propanol ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Neopentyl alcohol

D0792-25G D0792-500G D0792-100G
TCIAD0792-25GEA 2525 NOK
TCIAD0792-25G TCIAD0792-500G TCIAD0792-100G
2,2-Dimetyl-1-propanol ≥98.0% (by GC)
2,2-Dimetyl-1-propanol
Formel: C₅H₁₂O
MW: 88.15 g/mol
Kokepunkt: 113
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004682
CAS-nummer: 75-84-3
EINECS: 200-907-3
FN: 1325
ADR: 4.1 III
Merck Indeks: 14,06457
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9061f0a5a7fc01a09802ded111e34927eee5e23e&vid=9061f0a5a7fc01a09802ded111e34927eee5e23e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 53.0 to 57.0 deg-C
Solubility in Methanol almost transparency

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR