tert-Butanol, vannfri ≥99.5%
Leverandør: Thermo Scientific
|
Danger
|
Packaged under argon in resealable ChemSeal™ bottles.
41857.AK
41857.K2
41857.M1
41857.AD
41857.AKEA
870
NOK
41857.AK
41857.K2
41857.M1
41857.AD
tert-Butanol, vannfri ≥99.5%
tert-Butanol
Purity: 99.5%
CAS No.: 75-65-0
Molecular Formula: C4H10O
Formula Weight: 74.12
Flash Point: 11°(52°F)
Physical Form: Liquid
Density: 0.786
Refractive Index: 1.3870
Appearance: Colourless
Melting Point: 25°
Boiling Point: 83°
Beilstein Reference: 906698
Merck Reference: 14,1542
UN No.: 1120
MDL No.: MFCD00004464
Formel:
(CH₃)₃COH Kokepunkt: 83 °C Smeltepunkt: 25 °C Tetthet: 0,786 Flash punkt: 11 °C (52 °F) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00004464 CAS-nummer: 75-65-0 EINECS: 200-889-7 FN: 1120 ADR: 3,II Merck Indeks: 14,1542 |