Homovanillic acid ≥98%
Leverandør: Thermo Scientific
Warning
Synonymer: 2-(4-Hydroxy-3-methoxyphenyl)acetic acid, (4-Hydroxy-3-methoxyphenyl)acetic acid
A12441.03 A12441.14
A12441.03EA 905 NOK
A12441.03 A12441.14
Homovanillic acid ≥98%
Homovanillic acid
Purity: 98+% CAS No.: 306-08-1 Molecular Formula: C9H10O4 Formula Weight: 182.18 Melting Point: 140-145° Beilstein Reference: 2213447 Merck Reference: 14,4740 MDL No.: MFCD00004350
Formel: (C₆H₃(OH)(OCH₃))CH₂COOH
MW: 182,18 g/mol
Kokepunkt: 339 °C (1013 hPa)
Smeltepunkt: 140…145 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004350
CAS-nummer: 306-08-1
EINECS: 206-176-7
Merck Indeks: 14,4740