HEPES sodium salt ≥99%
Leverandør: Thermo Scientific
Synonymer: Sodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulphonate, Sodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulphonate, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid) sodium salt, 4-(2-Hydroxyethyl)piperazine-1-ethanesulphonic acid sodium salt, HEPES sodium, HEPES Na
A16516.14 A16516.22
A16516.14EA 665 NOK
A16516.14 A16516.22
HEPES sodium salt ≥99%
HEPES sodium salt
Purity: 99% CAS No.: 75277-39-3 Molecular Formula: C8H17N2NaO4S Formula Weight: 260.28 MDL No.: MFCD00036463
Formel: C₈H₁₇N₂NaO₄S
MW: 260,29 g/mol
Smeltepunkt: ? 105 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00036463
CAS-nummer: 75277-39-3
EINECS: 278-169-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=116e9b4f3c17d013ff2b7c9848c219943a725b3c&vid=8c800de68c4434c3c9d77107ff6779ef516bd851&prid=web&clid=SAPDAM