4-Methylcinnamic acid predominantly trans 99%
Leverandør: Thermo Scientific
Warning
Synonymer: 3-(p-Tolyl)acrylic acid
A13337.06 A13337.14 A13337.22
A13337.06EA 445 NOK
A13337.06 A13337.14 A13337.22
4-Methylcinnamic acid predominantly trans 99%
4-Methylcinnamic acid
Purity: 99% CAS No.: 1866-39-3 Molecular Formula: C10H10O2 Formula Weight: 162.19 Melting Point: 196-200° Beilstein Reference: 1364327
Formel: C₁₀H₁₀O₂
MW: 162,19 g/mol
Kokepunkt: 303 °C (1013 hPa)
Smeltepunkt: 196…200 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002697
CAS-nummer: 1866-39-3
EINECS: 217-479-9