3-(Trifluoromethyl)cinnamic acid predominantly trans ≥98%
Leverandør: Thermo Scientific
Warning
Synonymer: 3-(3-(Trifluoromethyl)phenyl)acrylic acid
A16983.06 A16983.14
A16983.06EA 640 NOK
A16983.06 A16983.14
3-(Trifluoromethyl)cinnamic acid predominantly trans ≥98%
3-(Trifluoromethyl)cinnamic acid
Purity: 98+% CAS No.: 779-89-5 Molecular Formula: C10H7F3O2 Formula Weight: 216.16 Odour: Odourless Melting Point: 132-136° Beilstein Reference: 1963372 MDL No.: MFCD00004393
Formel: C₁₀H₇F₃O₂
MW: 216,16 g/mol
Kokepunkt: 292 °C (1013 hPa)
Smeltepunkt: 132…136 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004393
CAS-nummer: 779-89-5
EINECS: 212-301-6