Sinapic acid predominantly trans 98%
Leverandør: Thermo Scientific
Warning
Synonymer: 3,5-Dimethoxy-4-hydroxycinnamic acid, 3',5'-Dimethoxy-4'-hydroxycinnamic acid, 4-Hydroxy-3,5-dimethoxycinnamic acid
115550010 115550050 115550250
ACRO115550010EA 560 NOK
ACRO115550010 ACRO115550050 ACRO115550250
Sinapic acid predominantly trans 98%
Sinapic acid
Formel: C₁₁H₁₂O₅
MW: 224,21 g/mol
Kokepunkt: 378 °C (1013 hPa)
Smeltepunkt: 203…205 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004401
CAS-nummer: 530-59-6
EINECS: 208-487-3
Bestill nå
Spesifikasjon Testresultater
Appearance Yellow to light beige Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %