o-Acetylsalisylsyre ≥99%
Leverandør: Thermo Scientific
Warning
Synonymer: 2-Acetoxybenzoic acid, Acetylsalicylic acid, Aspirin, Aspirin®
A12488.0E A12488.22 A12488.36
A12488.0EEA 2525 NOK
A12488.0E A12488.22 A12488.36
o-Acetylsalisylsyre ≥99%
o-Acetylsalicylic acid
Purity: 99% CAS No.: 50-78-2 Molecular Formula: C9H8O4 Formula Weight: 180.15 Flash Point: 250°(482°F) Melting Point: 133-140° Beilstein Reference: 779271 Merck Reference: 14,851 MDL No.: MFCD00002430
Formel: 2-(CH₃CO₂)C₆H₄CO₂H
MW: 180,16 g/mol
Kokepunkt: 140 °C (1013 hPa)
Smeltepunkt: 133…140 °C
Tetthet: 1,400
Flash punkt: 250 °C (482 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002430
CAS-nummer: 50-78-2
EINECS: 200-064-1
Merck Indeks: 14,851
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fcb60a4ee505b596cef38216a85e034985091e0&vid=5f8d5cfeb3039783869a9320ec840b8ec12c33ea&prid=web&clid=SAPDAM