1,2-Difluoro-4-nitrobenzene ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
3,4-Difluoronitrobenzene
D1926-250G
D1926-25G
TCIAD1926-250GEA
2725
NOK
TCIAD1926-250G
TCIAD1926-25G
1,2-Difluoro-4-nitrobenzene ≥98.0% (by GC)
1,2-Difluoro-4-nitrobenzene
Formel:
C₆H₃F₂NO₂ MW: 159,09 g/mol Kokepunkt: 78°C Tetthet: 1,43 g/cm³ (20 °C) Flash punkt: 80°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00007198 CAS-nummer: 369-34-6 EINECS: 206-718-2 ADR: 6.1,III |
Spesifikasjon Testresultater
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...