Phenylacetylene for syntese, Sigma-Aldrich®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

8.07044.0025 8.07044.0100
8.07044.0025EA 471 NOK
8.07044.0025 8.07044.0100
Phenylacetylene for syntese, Sigma-Aldrich®
Phenylacetylene
Formel: C₈H₆
MW: 102,14 g/mol
Kokepunkt: 144 °C (1013 hPa)
Smeltepunkt: –44,8 °C
Tetthet: 1,1 g/cm³ (20 °C)
Flash punkt: 31 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00008570
CAS-nummer: 536-74-3
EINECS: 208-645-1
FN: 1993
ADR: 3,III
Merck Indeks: 13,03892

Bestill nå

Spesifikasjon Testresultater

Solubility 0.46 g/l (25 °C)
Melting Point -44.8 °C
Molar Mass 102.13 g/mol
Boiling Point 142 - 144 °C (1013 hPa)
Vapor Pressure 2.7 hPa (25 °C)
Flash Point 27 °C
Refractive Index 1.547 (20 °C, 589 nm)
Density 0.93 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR