2-Bromobenzyl alcohol ≥98%
Leverandør: Thermo Scientific
Synonymer: (2-Bromophenyl)methanol
A15700.09 A15700.18
A15700.09EA 540 NOK
A15700.09 A15700.18
2-Bromobenzyl alcohol ≥98%
2-Bromobenzyl alcohol
Purity: 98% CAS No.: 18982-54-2 Molecular Formula: C7H7BrO Formula Weight: 187.04 Melting Point: 78-82° Beilstein Reference: 2243418
Formel: C₇H₇BrO
MW: 187,04 g/mol
Kokepunkt: 255 °C (1013 hPa)
Smeltepunkt: 78…82 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004600
CAS-nummer: 18982-54-2
EINECS: 242-719-4