3-Nitro-p-toluic acid ≥99.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
4-Methyl-3-nitrobenzoic acid
M0950-25G
M0950-500G
TCIAM0950-25GEA
230
NOK
TCIAM0950-25G
TCIAM0950-500G
3-Nitro-p-toluic acid ≥99.0% (by titrimetric analysis)
3-Nitro-p-toluic acid
Formel:
C₈H₇NO₄ MW: 181,15 g/mol Kokepunkt: 360 °C (1013 hPa) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00007174 CAS-nummer: 96-98-0 EINECS: 202-549-3 |
Spesifikasjon Testresultater
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 188.0 to 192.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...