Bromkresolgrønt ACS
Leverandør: Thermo Scientific
Synonymer: 3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
402930010 402930100 402930050
ACRO402930010EA 380 NOK
ACRO402930010 ACRO402930100 ACRO402930050
Bromkresolgrønt ACS
Bromkresolgrønt
Formel: C₂₁H₁₄Br₄O₅S
MW: 698,02 g/mol
Smeltepunkt: 225 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005874
CAS-nummer: 76-60-8
EINECS: 200-972-8
Merck Indeks: 13,01370
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12b4d4b3d992f8e86746537bf756af442752c853&vid=553ef1836a5b191a43558f8550e78cf2e424446e&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Yellow to beige to light brown Powder
Infrared spectrum Conforms
Visual transition interval from pH 3.8 (yellow) to pH 5.4 (blue)
Clarity of solution Passes test