Bly(II)nitrat ≥99%
Leverandør: Thermo Scientific
Danger
Synonymer: Lead dinitrate
A16345.0E A16345.22 A16345.36 A16345.PG
A16345.0EEA 2925 NOK
A16345.0E A16345.22 A16345.36 A16345.PG
Bly(II)nitrat ≥99%
Bly(II)nitrat
Purity: 99% CAS No.: 10099-74-8 Molecular Formula: Pb(NO3)2 Formula Weight: 331.20 Odour: Odourless Melting Point: 470° UN No.: 1469 MDL No.: MFCD00011153
Formel: Pb(NO₃)₂
MW: 331,21 g/mol
Smeltepunkt: 470 °C
Tetthet: 4,530
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011153
CAS-nummer: 10099-74-8
EINECS: 233-245-9
FN: 1469
ADR: 5.1,II
Merck Indeks: 14,5414