Triphenyl phosphite for syntese, Sigma-Aldrich®
Leverandør: Merck
Warning
M-Clarity™ quality level = MQ200
8.00551.0100 8.00551.1000
8.00551.0100EA 363 NOK
8.00551.0100 8.00551.1000
Triphenyl phosphite for syntese, Sigma-Aldrich®
Triphenyl phosphite
Formel: C₁₈H₁₅O₃P
MW: 310,29 g/mol
Kokepunkt: 360 °C (1013 hPa)
Smeltepunkt: 22…25 °C
Tetthet: 1,19 g/cm³ (20 °C)
Flash punkt: 188 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003032
CAS-nummer: 101-02-0
EINECS: 202-908-4
FN: 3077
ADR: 9,III
Bestill nå
Spesifikasjon Testresultater
Solubility (20 °C) (slow decomposition)
Melting Point 22 - 25 °C
Molar Mass 310.29 g/mol
Boiling Point 360 °C (1013 hPa) decomposes
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 188 °C
Refractive Index 1.5903 (20 °C, 589 nm)
Density 1.19 g/cm3 (20 °C)
pH 1 (200 g/l, H2O, 20 °C)