(1-Dodecyl)triphenylphosphonium bromide ≥98%
Leverandør: Thermo Scientific
Warning
Synonymer: Dodecyltriphenylphosphonium bromide
A14295.09 A14295.18 A14295.30
A14295.09EA 980 NOK
A14295.09 A14295.18 A14295.30
(1-Dodecyl)triphenylphosphonium bromide ≥98%
(1-Dodecyl)triphenylphosphonium bromide
Purity: 98+% CAS No.: 15510-55-1 Molecular Formula: CH3(CH2)11P(C6H5)3Br Formula Weight: 511.54 Melting Point: 90-94° Beilstein Reference: 3581921
Formel: H₃C(CH₂)₁₁P(C₆H₅)₃Br
MW: 511,53 g/mol
Smeltepunkt: 90…94 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00031546
CAS-nummer: 15510-55-1
EINECS: 239-538-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0fe7db87e7b9e3a4f94baa8840868d0edbeaaa6&vid=e0fe7db87e7b9e3a4f94baa8840868d0edbeaaa6&prid=web&clid=SAPDAM