5-Iodoindole ≥98%
Leverandør: Thermo Scientific
|
Warning
|
B20638.06
B20638.14
B20638.06EA
1350
NOK
B20638.06
B20638.14
5-Iodoindole ≥98%
5-Iodoindole
Purity: 98%
CAS No.: 16066-91-4
Molecular Formula: C8H6IN
Formula Weight: 243.05
Melting Point: 97-100°
MDL No.: MFCD00220065
Formel:
C₈H₆IN MW: 243,05 g/mol Smeltepunkt: 97…100 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00220065 CAS-nummer: 16066-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...