1-Naphthoic acid ≥98%
Leverandør: Thermo Scientific
Warning
Synonymer: α-Naphthoic acid
A10273.14 A10273.22
A10273.14EA 460 NOK
A10273.14 A10273.22
1-Naphthoic acid ≥98%
1-Naphthoic acid
Purity: 98% CAS No.: 86-55-5 Molecular Formula: C11H8O2 Formula Weight: 172.18 Flash Point: 195°(383°F) Melting Point: 160-162° Boiling Point: 300° Beilstein Reference: 1908896 Merck Reference: 14,6381 MDL No.: MFCD00004007
Formel: C₁₁H₈O₂
MW: 172,18 g/mol
Kokepunkt: >300 °C
Smeltepunkt: 160…162 °C
Tetthet: 1,398
Flash punkt: 195 °C (383 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004007
CAS-nummer: 86-55-5
EINECS: 201-681-9
Merck Indeks: 14,6381