Zinkoksid, HiFUEL® A310 for sulfur removal
Leverandør: Thermo Scientific
Warning
Synonymer: Zinci oxidum, Zinc (II) oxide
45469.A4 45469.A1 45469.36
45469.A4EA 6725 NOK
45469.A4 45469.A1 45469.36
Zinkoksid, HiFUEL® A310 for sulfur removal
Zinc oxide
Physical Form: Spheres, 2-5mm dia MDL No.: MFCD00011300
Formel: ZnO
MW: 81,39 g/mol
Smeltepunkt: 1975 °C
Tetthet: 5,61 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011300
CAS-nummer: 1314-13-2
EINECS: 215-222-5
FN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Indeks: 13,10200