Phenanthridine 98%
Leverandør: Thermo Scientific
|
Warning
|
169250050
169250010
169250250
ACRO169250050EA
1450
NOK
ACRO169250050
ACRO169250010
ACRO169250250
Phenanthridine 98%
Phenanthridine
Formel:
C₁₃H₉N MW: 179,22 g/mol Kokepunkt: 349 °C (769 mmHg) Smeltepunkt: 104…108 °C Tetthet: 1,1 g/cm³ (130 °C) Flash punkt: 100 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00004989 CAS-nummer: 229-87-8 EINECS: 205-934-4 ADR: 6.1,III |
Spesifikasjon Testresultater
Appearance | White to beige Powder |
Infrared spectrum | Conforms |
Melting point | 104 to 108 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...