4-Amino-2-chloropyridine 98%
Leverandør: Thermo Scientific
Warning
Synonymer: 2-Chloro-4-aminopyridine
B22098.06 B22098.14
B22098.06EA 990 NOK
B22098.06 B22098.14
4-Amino-2-chloropyridine 98%
4-Amino-2-chloropyridine
Purity: 98% CAS No.: 14432-12-3 Molecular Formula: C5H5ClN2 Formula Weight: 128.56 Odour: Unpleasant Melting Point: 91-94° Boiling Point: 153°/5mm Beilstein Reference: 108671 MDL No.: MFCD00060089
Formel: C₅H₅ClN₂
MW: 128,56 g/mol
Kokepunkt: 153 °C (5 mmHg)
Smeltepunkt: 91…94 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00060089
CAS-nummer: 14432-12-3
EINECS: 238-403-0