VWR® PerfectBlue Twin M, Dual-Gel System
Leverandør: VWR Collection
700-0955EA 28275 NOK
700-0955 700-0961
VWR® PerfectBlue Twin M, Dual-Gel System
Elektroforesesystemer Vertikal gelelektroforesesystemer
For 16×14 cm gels, the Twin M is a versatile system for SDS PAGE applications, in addition to non denaturing electrophoresis of proteins, SSCPs and dinucleotide repeats. With the enlarged upper buffer chamber, water circulation system and heat-sinking aluminium ceramic plates, the Twin M provides high speed separation while maintaining straight, sharp bands.
*Based on an average gel and a tooth at 20 mm depth
Bestillingsinformasjon: Complete system, with or without casting base, includes internal cooling core, SuperSafe cover with attached leads, 4× glass plate pair sets, 4× combs (1,5More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Buffervolum, geltank ~650 ml
Prøvekapasitet 10 – 48
Enhet, B× H × D 250×140×200 mm