VWR® PerfectBlue Twin S, Dual-Gel System
Leverandør: VWR Collection
700-0931EA 20775 NOK
700-0931 700-0937
VWR® PerfectBlue Twin S, Dual-Gel System
Elektroforesesystemer Vertikal gelelektroforesesystemer
The PerfectBlue Twin S system is designed for rapid separation of typical 10×10 cm mini gels. Due to its small size and a separation capacity of 6 to 30 samples, it is ideal for fast, analytical separations. A highly efficient cooling system ensures uncompromising results.
*Based on an average gel and a tooth with 14 mm depth.
Bestillingsinformasjon: Complete system, with or without casting base, includes internal cooling core, SuperSafe cover with attached leads, 4× glass plate pair sets, 2× combs (0,8 mm thick, 10 teeth), 2× spacer sets (0,8 mm), 2× replacement gaskets, 1× blockingMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Buffervolum, geltank ~250 ml
Prøvekapasitet 6 – 24
Enhet, B× H × D 170×130×140 mm