VWR® PerfectBlue Twin ExW S, Dual-Gel System
Leverandør: VWR Collection
700-0980EA 28400 NOK
700-0980 700-0986
VWR® PerfectBlue Twin ExW S, Dual-Gel System
Elektroforesesystemer Vertikal gelelektroforesesystemer
Extra wide, for gels 20×10 cm, Twin ExW S is ideally suited to rapid analytical separations or large sample separation. The combination of the triple-cathode construction, and effective cooling provides a stable environment for high quality sample separation.
*Based on an average gel and a tooth with 14 mm depth.
Bestillingsinformasjon: Complete system, with or without casting base, includes internal cooling core, SuperSafe cover with attached leads, 4× glass plate pair sets, 4× combs (0,8 mm thick, 2×15 teeth and 2×20 teeth), 4× spacer sets (0,8 mm), 2× replacement gaskets, 1×More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Buffervolum, geltank ~650 ml
Prøvekapasitet 10 − 72
Enhet, B× H × D 300×130×140 mm