Syringe filters, colour coded
Leverandør: MICROSOLV
58045-P25-C 58045-R25-C 58045-N25-C
MICS58045-P25-CEA 3000 NOK
MICS58045-P25-C MICS58045-R25-C MICS58045-N25-C
Syringe filters, colour coded
Filter Sprøytefilter
Colour coded low volume syringe filter for HPLC, CE or dissolution testing.
  • Hydrophilic membrane
  • Houding made from virgin medical grade PP
  • Retention volume of 90 µl
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Maks. bemanningstrykk 7,6 bar
Maks. temp omgivelser 100 °C