Aluminiumoksid grunn 32-63 µm APS HPLC flash grade
Leverandør: Thermo Scientific
46071.22 46071.36
46071.22EA 460 NOK
46071.22 46071.36
Aluminiumoksid grunn 32-63 µm APS HPLC flash grade
Aluminium oxide basic
Formel: Al₂O₃
MW: 101,96 g/mol
MDL nummer: MFCD00003424
CAS-nummer: 1344-28-1