ESD safety shoes, lace-up, Safety-Grip, Till
Leverandør: ELTEN GMBH
1721191/36 1721191/37 1721191/38 721191/39 721191/40 721191/41 721191/42 721191/43 721191/44 721191/45 721191/46 721191/47 721191/48
ELTN1721191/36EA 1025 NOK
ELTN1721191/36 ELTN1721191/37 ELTN1721191/38 ELTN721191/39 ELTN721191/40 ELTN721191/41 ELTN721191/42 ELTN721191/43 ELTN721191/44 ELTN721191/45 ELTN721191/46 ELTN721191/47 ELTN721191/48
ESD safety shoes, lace-up, Safety-Grip, Till
Sko og støvler
ESD safety shoes made from water resistant cowhide leather upper, a full padded bellows tongue plus a slip resistant, dual-density rubber/PU sole (Safety-Grip) (3,5 mm) with exceptional non slip and good self-cleaning properties.
  • Climate-regulating, breathable textile lining
  • Full length fleece inlay sole (ESD Pro), black
  • Replaceable insole; orthopaedic insoles possible
  • Soft fleece ESD insole
  • Anatomically-shaped insole and ankle support for improved stability
  • TPU scuff cap protects the upper
  • Puncture resistant steel midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Sertifiseringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC, HI, HRO