Propylphosphonic acid ≥98%, crystalline solid

Leverandør: Cayman Chemical
Danger

21702-1 21702-10 21702-25 21702-5
CAYM21702-1EA 1400 NOK
CAYM21702-1 CAYM21702-10 CAYM21702-25 CAYM21702-5
Propylphosphonic acid ≥98%, crystalline solid
Propylphosphonic acid

Propylphosphonic acid is a compound that is highly susceptible to degradation by radical species. It is used to investigate the inactivation of A. hydrophila by free radicals produced spontaneously in oxidizing iron-containing groundwaters and also to to identify toxic nerve agents in water samples.


NB: Not for human or veterinary use.

Formel: C₃H₉O₃P
Lagringstemperatur: Fryseskap
CAS-nummer: 4672-38-2
FN: 3260
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Purity ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR