SPE columns, CHROMABOND® HILIC
Leverandør: MACHEREY-NAGEL
730593 730594
MANA730593EA 5475 NOK
MANA730593 MANA730594
SPE columns, CHROMABOND® HILIC
Kromatografikolonner
CHROMABOND® HILIC is an ammonium – sulfonic acid modified silica phase for SPE.
  • Zwitterionic polar phase
  • Hydrophilic interaction chromatography (HILIC)
  • Polar analytes are more strongly retained than nonpolar compounds
The orthogonal behaviour of CHROMABOND® HILIC to RP materials shows good recoveries for an extraction of polar organic acids and bases, polar natural compounds, nucleosides, oligonucleotides, amino acids, peptides, and water-soluble vitamins.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Karbonbelastning 7%
pH område 2 - 8
Overflate areal 500 m²/g