Perchloric acid concentrate for dilution to 1L, 0.1 M HClO4 (0.1N) volumetrisk løsning, for titration in non-aqueous media, Fluka™
Leverandør: Honeywell Chemicals
Danger
32046-6X1EA 32046-1EA
32046-6X1EAEA 6175 NOK
32046-6X1EA 32046-1EA
Perchloric acid concentrate for dilution to 1L, 0.1 M HClO4 (0.1N) volumetrisk løsning, for titration in non-aqueous media, Fluka™
Perchloric acid
Concentrate for 1L standard solution
Formel: HClO₄
Kokepunkt: 203 °C
Smeltepunkt: -18 °C
Tetthet: 1,66 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011325
CAS-nummer: 7601-90-3
FN: 1873
ADR: 5.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Factor 0.998 - 1.002