Strips of 8 PCR tubes with attached flat individual caps
Leverandør: Brand
781316 781317 781315 781318
732-1504EA 3500 NOK
732-1504 732-1505 732-3613 732-3614
Strips of 8 PCR tubes with attached flat individual caps
Rør PCR-rør
PP. Eight connected 0,2 or 0,15 ml PCR tubes with attached flat individual caps. The tubes are held together by three connectors for increased rigidity.
  • Cap offers protection against contamination
  • Highly transparent cap makes them ideal for real-time PCR (qPCR)
  • Available in standard or low profile versions
  • Free from DNase, RNase and DNA
Colour: Clear or white