BMS 195614 ≥98%
Leverandør: Cayman Chemical
Synonymer: BMS-195614, BMS195614, BMS 614, BMS-614, BMS614
16029-1 16029-10 16029-5
CAYM16029-1EA 995 NOK
CAYM16029-1 CAYM16029-10 CAYM16029-5
BMS 195614 ≥98%
BMS 195614
BMS 195614 is a neutral retinoic acid receptor (RAR) α-selective antagonist (Ki = 2.5 nM). It antagonizes agonist-induced coactivator recruitment and moderately decreases SMRT binding to RAR but does not significantly affect nuclear receptor corepressor binding.
  • Formulation: A crystalline solid
  • λmax: 211, 236, 282 nm
  • Storage -20°C
Formel: C₂₉H₂₄N₂O₃
MW: 448,5 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00952209
CAS-nummer: 182135-66-6