VWR®, Condensers with Cooling Coil and Jacket
Leverandør: VWR Collection
201-1416EA 1150 NOK
201-1416 201-1417 201-1405 201-1408
VWR®, Condensers with Cooling Coil and Jacket
Kondensator
Borosilicate 3.3 glass.
  • Very good chemical resistance
  • High temperature resistance and high resistance to thermal shock
  • Standard ground cone and socket
These condensers, with cooling coil and jacket, have an additional cooling surface for condensing highly volatile vapours. With GL 14 screw thread and 8 mm external diameter tubing connection.