Supelco® Organic Reference Standard, Isovaleryl-ʟ-carnitine-(N,N,N-trimethyl-d₉)
Leverandør: Merck
52993-10MG 52993-5MG
52993-10MGEA 3850 NOK
52993-10MG 52993-5MG
Supelco® Organic Reference Standard, Isovaleryl-ʟ-carnitine-(N,N,N-trimethyl-d₉)
Standarder Organiske standarder
  • GHS signal: GHS07
NB: P264 - P280 - P302 + P352 - P305 + P351 + P338 - P332 + P313 - P337 + P313
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb057cae6af233e2d1979d3d46aa89c74557d73c&vid=4d74b2d20129ac5f503620de21072ddd749185e6&prid=web&clid=SAPDAM
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
M-Clarity quality level MQ100
Synonym(s) (2R)-3-Carboxy-N,N,N-tri(methyl-d₃)-2-(3-methyl-1-oxobutoxy)-1-propanaminium inner salt; ʟ-Carnitine-(trimethyl-d₉) isovaleryl ester
CAS-nr 203806-03-5
MDL No. MFCD01073493
Molekylvekt 254,37
Hardhetsklasse beskrivelse H315 - H319
Fraktforhold WICE
Lagringsforhold –20 °C