Exsil ODS, HPLC Columns
Leverandør: Avantor
EXODS1-5-250AEA 6175 NOK
EXODS1-5-250A EXODS-10-250P EXODS-5-100AM EXODS-5-10CP3 EXODS-5-250AF EXODS-5-250P
Exsil ODS, HPLC Columns
Kromatografikolonner
Exsil HPLC columns are a useful alternative to the classical spherical HPLC silicas.
  • Spherical porous silica
  • Competitively priced
  • Hichrom high efficiency
Exsil 100, the standard material, is supplied in a range of particle sizes and bonded phases to cover a wide range of applications.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Karbonbelastning 7 11 11 11
Kappet ved endene Nei Yes Ja Ja
Overflate areal 200 m²/g 200 m²/g 200 m²/g 200 m2 /g
Miljøsertifikat ISO 14001:2015 (530840 UM15) ISO 14001:2015 (530840 UM15) ISO 14001:2015 (530840 UM15)