Superspher® RP18, HPLC Columns
Leverandør: Avantor
SUPERRP18-4-125AFEA 11325 NOK
SUPERRP18-4-125AF SUPERRP184250A SUPERRP18E475A SUPERRP18425AF HICHSUPERRP184125M
Superspher® RP18, HPLC Columns
Kromatografikolonner
A range of 4 µm materials has been marketed under the Superspher® tradename in Europe.
  • Spherical porous silica
  • Base deactivated LiChrospher RP-select B
  • Endcapping options for RP-8 and RP-18
  • Hichrom high efficiency packed columns
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Karbonbelastning 21,0%
Kappet ved endene Nei
Overflate areal 350 m²/g