Ammoniumacetat, LiChropur™ for HPLC, Supelco®
Leverandør: Merck
Synonymer: Acetic acid ammonium salt
M-Clarity™ quality level = MQ100
5.43834.0100 5.43834.0250
5.43834.0100EA 890 NOK
5.43834.0100 5.43834.0250
Ammoniumacetat, LiChropur™ for HPLC, Supelco®
Ammoniumacetat
Formel: H₃CCOONH₄
MW: 77.08 g/mol
Smeltepunkt: 114 °C
Tetthet: 1.17 g/cm³ (20 °C)
Flash punkt: 136
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013066
CAS-nummer: 631-61-8
EINECS: 211-162-9
Merck Indeks: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM
Limited Time Sale
Select items below are currently on sale